Learn More
Aflatoxin G2-d3, TRC
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
Specifications
Specifications
| Chemical Name or Material | Aflatoxin G2-d3 |
| CAS | 1217758-21-8 |
| Molecular Formula | C17 D3 H11 O7 |
| Quantity | 0.5 mg |
| Synonym | (7aR,10aS)-3,4,7a,9,10,10a-Hexahydro-5-(methoxy-d3)-1H,12H-furo[3',2':4,5]furo[2,3-h]pyrano[3,4-c][1]benzopyran-1,12-dione, Dihydroaflatoxin G1-d3 |
| InChI Formula | InChI=1S/C17H14O7/c1-20-9-6-10-12(8-3-5-22-17(8)23-10)14-11(9)7-2-4-21-15(18)13(7)16(19)24-14/h6,8,17H,2-5H2,1H3/t8-,17+/m0/s1/i1D3 |
| SMILES | [2H]C([2H])([2H])Oc1cc2O[C@H]3OCC[C@H]3c2c4OC(=O)C5=C(CCOC5=O)c14 |
| IUPAC Name | (3S,7R)-11-(trideuteriomethoxy)-6,8,16,20-tetraoxapentacyclo[10.8.0.02,9.03,7.013,18]icosa-1,9,11,13(18)-tetraene-17,19-dione |
| Molecular Weight (g/mol) | 333.31 |
| Formula Weight | 333.0928 |