missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atovaquone-d5, TRC
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
Specifications
Specifications
| Chemical Name or Material | Atovaquone-d5 |
| CAS | 1329792-63-3 |
| Molecular Formula | C22 D5 H14 Cl O3 |
| Quantity | 0.5 mg |
| Synonym | 2-[trans-4-(4-Chlorophenyl)cyclohexyl-d5-]-3-hydroxy-1,4-naphthalenedione, 566C80-d5, BW-566C-d5, BW-566C-80-d5, Mepron-d5, Wellvone-d5 |
| InChI Formula | InChI=1S/C22H19ClO3/c23-16-11-9-14(10-12-16)13-5-7-15(8-6-13)19-20(24)17-3-1-2-4-18(17)21(25)22(19)26/h1-4,9-13,15,26H,5-8H2/t13?,15-/i7D2,8D2,15D |
| SMILES | [2H]C1([2H])CC(CC([2H])([2H])C1([2H])C2=C(O)C(=O)c3ccccc3C2=O)c4ccc(Cl)cc4 |
| IUPAC Name | 2-[4-(4-chlorophenyl)-1,2,2,6,6-pentadeuterio-cyclohexyl]-3-hydroxy-naphthalene-1,4-dione |
| Molecular Weight (g/mol) | 371.87 |
| Formula Weight | 371.1337 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction