missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Bis(di-tert-butylphosphino)ferrocene palladium dichloride, Palladium (Pd) 15.8, Thermo Scientific Chemicals

Catalog Number p-7077401
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
95408-45-0
C26H44Cl2FeP2Pd
651.75
MFCD08064219
DFRHTHSZMBROSH-UHFFFAOYSA-N
Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II)
[Fe].Cl[Pd]Cl.CC(C)(C)P(c1cccc1)C(C)(C)C.CC(C)(C)P(c1cccc1)C(C)(C)C
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
95408-45-0
C26H44Cl2FeP2Pd
651.75
MFCD08064219
DFRHTHSZMBROSH-UHFFFAOYSA-N
Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II)
[Fe].Cl[Pd]Cl.CC(C)(C)P(c1cccc1)C(C)(C)C.CC(C)(C)P(c1cccc1)C(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Infrared Spectrum Authentic
Quantity 250 mg
Solubility Information Solubility in water: insoluble.
Formula Weight 651.75
Percent Purity 10.1 to 11.6% (Cl) (Argentometry)
Chemical Name or Material 1,1'-Bis(di-tert-butylphosphino)ferrocene palladium dichloride
compliance-icons
Product Identifier
  • 1,1'-Bis(di-tert-butylphosphino)ferrocene palladium dichloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H413-May cause long lasting harmful effects to aquatic life.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'