missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Dimethyl-4,4'-bipyridinium Dichloride, TRC

Product Code. 30374407
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30374407

Brand: Toronto Research Chemicals TRCD472695100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Paraquat dichloride
CAS 1910-42-5
Molecular Formula C12 H14 N2 . 2 Cl
Quantity 100 mg
Synonym 4,4'-Bipyridinium, 1,1'-dimethyl-, chloride (1:2), 1,1'-Dimethyl-4,4'-bipyridinium dichloride (6CI,7CI), 4,4'-Bipyridinium, 1,1'-dimethyl-, dichloride (8CI,9CI), 1,1'-Dimethyl-4,4'-dipyridinium dichloride, 1,1'-Dimethyl-4,4'-dipyridylium dichloride, AH 501, AH 501 (herbicide), Cekuquat, Cyclone Max, Dimethyl viologen chloride, Galokson, Gramixel, Gramoxone, Gramoxone D, Gramoxone Inteon, Gramoxone Max, Gramoxone S, Gramoxone W, Gramoxone dichloride, Methyl viologen, Methyl viologen dichloride, Methylviologen chloride, N,N'-Dimethyl-4,4'-bipyridinium dichloride, N,N'-Dimethyl-4,4'-dipyridylium dichloride, Nuquat, OK 622, Paragreen A, Paraquat, Paraquat dichloride
InChI Formula InChI=1S/C12H14N2.2ClH/c1-13-7-3-11(4-8-13)12-5-9-14(2)10-6-12;;/h3-10H,1-2H3;2*1H/q+2;;/p-2
SMILES [Cl-].[Cl-].C[n+]1ccc(cc1)c2cc[n+](C)cc2
IUPAC Name 1-methyl-4-(1-methylpyridin-1-ium-4-yl)pyridin-1-ium;dichloride
Molecular Weight (g/mol) 257.16
Formula Weight 256.0534
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.