Learn More
1,10-Decanediol, 97%, Thermo Scientific Chemicals
Catalog Number p-7021543
24.93 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
112-47-0
C10H22O2
174.284
MFCD00004749
FOTKYAAJKYLFFN-UHFFFAOYSA-N
1,10-decanediol, decamethylene glycol, decamethylenediol, 1,10-dihydroxydecane, 1,10-decamethylenediol, 1,10-decamethylene diol, 1,6-bis 2-hydroxyethyl hexane, alpha,omega-decanediol, unii-5i577udk52, 1,10-ddecanediol
37153
decane-1,10-diol
C(CCCCCO)CCCCO
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
112-47-0
C10H22O2
174.284
MFCD00004749
FOTKYAAJKYLFFN-UHFFFAOYSA-N
1,10-decanediol, decamethylene glycol, decamethylenediol, 1,10-dihydroxydecane, 1,10-decamethylenediol, 1,10-decamethylene diol, 1,6-bis 2-hydroxyethyl hexane, alpha,omega-decanediol, unii-5i577udk52, 1,10-ddecanediol
37153
decane-1,10-diol
C(CCCCCO)CCCCO
1,10-Decanediol is an minor intermediate in the production of polyesters. It also used in essence, perfumes and in pharmaceuticals. The diacrylate is hydrophobic and used for flexible dental materials, radiation curable coatings, and for lenses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,10-Decanediol is an minor intermediate in the production of polyesters. It also used in essence, perfumes and in pharmaceuticals. The diacrylate is hydrophobic and used for flexible dental materials, radiation curable coatings, and for lenses.
Solubility
Soluble in alcohol. Insoluble in water, petroleum ether.
Notes
Store in a cool, dry, well-ventilated area. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 71°C to 74°C
Density 1.08
Boiling Point 297°C
Flash Point 152°C (306°F)
Quantity 100 g
Beilstein 1698975
Merck Index 14,2849
Solubility Information Soluble in alcohol. Insoluble in water,petroleum ether.
Formula Weight 174.28
Percent Purity 97%
Chemical Name or Material 1,10-Decanediol