missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,10-Phenanthroline-5,6-dione, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
1,10-Phenanthroline-5,6-dione is a Bifunctional quinone oxidant which, when used in conjunction with Zn2+ catalysts, is used to affect the aerobic oxidation of secondary amines to a variety of value added motifs, including indoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,10-Phenanthroline-5,6-dione is a Bifunctional quinone oxidant which, when used in conjunction with Zn2+ catalysts, is used to affect the aerobic oxidation of secondary amines to a variety of value added motifs, including indoles.
Solubility
Insoluble in water.
Notes
Store in a cool and dark place. Store away from incompatible materials such as oxidizing agents.
1,10-Phenanthroline-5,6-dione is a Bifunctional quinone oxidant which, when used in conjunction with Zn2+ catalysts, is used to affect the aerobic oxidation of secondary amines to a variety of value added motifs, including indoles.
Solubility
Insoluble in water.
Notes
Store in a cool and dark place. Store away from incompatible materials such as oxidizing agents.
Specifications
Melting Point | 260°C (decomposition) |
Quantity | 250 mg |
Solubility Information | Insoluble in water. |
Formula Weight | 210.19 |
Percent Purity | 98% |
Chemical Name or Material | 1,10-Phenanthroline-5,6-dione |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'