Learn More
Trajan Scientific and Medical™ 1/16 in. Graphite/Vespel™ Fittings Reducing Ferrule
Made from 15% Graphite and 85% Vespel
Brand:  Trajan Scientific and Medical™ 0726632
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12786348
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • Composite of graphite and Vespel
  • Mechanically robust, hard material, long lifetime
  • Forms strong grip with capillary column
  • Hole I.D. smaller than nominal size of fitting
Specifications
Specifications
3.2 mm
3.7 mm
0.2 mm
15% Graphite, 85% Vespel
Graphite Vespel Reducing Ferrule 1/16 in. Fitting
Product Suggestions
Product Suggestions
Videos
SDS
Documenti
Documenti
Certificati
Offerte speciali
Offerte speciali