missing translation for 'onlineSavingsMsg'
Learn More

[1-[[[(1R)-1-[3-[(Z)-2-(7-Chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]sulfanyl]methyl]cyclopropyl]acetic Acid, Mikromol™

Product Code. 30050055
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30050055

Brand: Mikromol MM0698.130025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

[1-[[[(1R)-1-[3-[(Z)-2-(7-Chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]sulfanyl]methyl]cyclopropyl]acetic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material cis-Montelukast
CAS 774538-96-4
Analyte or Component Names cis-Montelukast
Shipping Condition Ice pack (-20°C)
Molecular Formula C35H36ClNO3S
Certifications/Compliance ISO 9001
Synonym 1-[[[(1R)-1-[3-[(1Z)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]thio]methyl]cyclopropaneacetic Acid, Montelukast Impurity G, Montelukast EP Impurity G, Montelukast USP Related Compound G, Montelukast cis Isomer
SMILES ClC1=CC=C(C=CC(/C=C\C2=CC=CC([C@H](SCC3(CC(O)=O)CC3)CCC4=C(C(C)(C)O)C=CC=C4)=C2)=N5)C5=C1
Molecular Weight (g/mol) 586.18
Quantity 25 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.