missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Benzenetricarboxylic Acid 1,2-Bis(2-ethylhexyl) Ester, TRC

Product Code. 30012580
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012580 10 mg 10mg
30138183 100 mg 100mg
30137980 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012580 Supplier Toronto Research Chemicals Supplier No. TRCB19003010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63468-09-7
Formula Weight 434.2668
InChI Formula InChI=1S/C25H38O6/c1-5-9-11-18(7-3)16-30-24(28)21-14-13-20(23(26)27)15-22(21)25(29)31-17-19(8-4)12-10-6-2/h13-15,18-19H,5-12,16-17H2,1-4H3,(H,26,27)
IUPAC Name 3,4-bis(2-ethylhexoxycarbonyl)benzoic acid
Molecular Formula C25 H38 O6
Molecular Weight (g/mol) 434.57
SMILES CCCCC(CC)COC(=O)c1ccc(cc1C(=O)OCC(CC)CCCC)C(=O)O
Chemical Name or Material 1,2,4-Benzenetricarboxylic Acid 1,2-Bis(2-ethylhexyl) Ester

Specifications

Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.