Learn More
1,2-Bis(2-nitrophenoxy)ethane, 98%, Thermo Scientific™
Product Code. 11470277
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. 11470277
Brand: Thermo Scientific Alfa Aesar B21692.09
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51661-19-9
Molecular Formula C14H12N2O6
Molecular Weight (g/mol) 304.26
MDL Number MFCD00024226
InChI Key DEHTVRKGDUCXRF-UHFFFAOYSA-N
Synonym 1,2-bis o-nitrophenoxy ethane, 1,2-bis 2-nitrophenoxy ethane, 1-nitro-2-2-2-nitrophenoxy ethoxy benzene, ethylene glycol bis 2-nitrophenyl ether, 1,1'-ethane-1,2-diylbis oxy bis 2-nitrobenzene, bis 2-nitrophenoxy ethane, acmc-20a59z, 1,2-di o-nitrophenoxy ethane, 2,2'-ethylenedioxy bis 1-nitrobenzene, 2-nitro-1-2-2-nitrophenoxy ethoxy benzene
PubChem CID 394580
IUPAC Name 1-nitro-2-[2-(2-nitrophenoxy)ethoxy]benzene
SMILES [O-][N+](=O)C1=CC=CC=C1OCCOC1=CC=CC=C1[N+]([O-])=O
Specifications
Melting Point 167°C to 170°C
Quantity 10 g
Formula Weight 304.26
Percent Purity 98%
Chemical Name or Material 1,2-Bis(2-nitrophenoxy)ethane