missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(dimethylphosphino)ethane, 97%, Thermo Scientific Chemicals

Product Code. 10482451 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10482451 500 mg 500mg
10580984 1 g 1g
10648254 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10482451 Supplier Thermo Scientific Acros Supplier No. 310035000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23936-60-9
Molecular Formula C6H16P2
Molecular Weight (g/mol) 150.14
MDL Number MFCD00008511
InChI Key ZKWQSBFSGZJNFP-UHFFFAOYSA-N
Synonym acmc-1cjp0, zkwqsbfsgzjnfp-uhfffaoysa, dimethyl-2-dimethylphosphino ethylphosphine, 1,2-bis dimethylphosphino ethane, 2-dimethylphosphanyl ethyl dimethylphosphane, 2-dimethylphosphanylethyl dimethyl phosphane, ethylenebis dimethylphosphine, pubchem6482, ghl.pd_mitscher_leg0.173, phosphine, 1,2-ethanediylbis dimethyl
PubChem CID 141059
SMILES CP(C)CCP(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.9 g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3)PCHCHP(CH3)
Refractive Index 1.5060 to 1.5080 (20°C, 589 nm)
Quantity 500 mg
IUPAC Name [2-(dimethylphosphanyl)ethyl]dimethylphosphane
Formula Weight 150.14
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 1, 2-Bis(dimethylphosphino)ethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.