missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloroisoquinoline, 97%, Thermo Scientific™

Product Code. 15476308
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15476308 1 g 1g
15486308 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15476308

Brand: Thermo Scientific Alfa Aesar H59873.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7742-73-6
Molecular Formula C9H5Cl2N
Molecular Weight (g/mol) 198.046
MDL Number MFCD00034750
InChI Key BRGZEQXWZWBPJH-UHFFFAOYSA-N
Synonym isoquinoline, 1,3-dichloro, 1,3-dichloro-isoquinoline, pubchem7254, 1, 3-dichloroisoquinoline, acmc-209p9x, ksc493q3b, 1,3-dichloroisoquinoline
PubChem CID 298625
IUPAC Name 1,3-dichloroisoquinoline
SMILES C1=CC=C2C(=C1)C=C(N=C2Cl)Cl

Specifications

Melting Point 121°C to 122°C
Quantity 1 g
Formula Weight 198.05
Percent Purity 97%
Chemical Name or Material 1,3-Dichloroisoquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.