missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diethylbenzene, 97+%, Thermo Scientific Chemicals

Catalog Number p-7058860
44.60 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
2 mL
10 mL
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
141-93-5
C10H14
134.222
MFCD00009260
AFZZYIJIWUTJFO-UHFFFAOYSA-N
m-diethylbenzene, benzene, 1,3-diethyl, benzene, m-diethyl, m-ethylethylbenzene, unii-zm2x7i1g8y, zm2x7i1g8y, ethylated benzene, benzene, ethylated, meta-diethylbenzene, polyethylated benzene
8864
1,3-diethylbenzene
CCC1=CC(=CC=C1)CC
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
141-93-5
C10H14
134.222
MFCD00009260
AFZZYIJIWUTJFO-UHFFFAOYSA-N
m-diethylbenzene, benzene, 1,3-diethyl, benzene, m-diethyl, m-ethylethylbenzene, unii-zm2x7i1g8y, zm2x7i1g8y, ethylated benzene, benzene, ethylated, meta-diethylbenzene, polyethylated benzene
8864
1,3-diethylbenzene
CCC1=CC(=CC=C1)CC

1,3-Diethylbenzene is used as an intermediate and solvent. It is also used in the preparation of divinyl benzene. Further, it is used in powderless etching.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.86 g/mL
Boiling Point 182°C
Flash Point 55°C (131°F)
Assay Percent Range >97%
Odor Aromatic
Refractive Index 1.496
Quantity 2 mL
UN Number UN2049
Solubility Information Miscible with ethanol,benzene,carbon tetrachloride,ethyl ether and acetone. Immiscible with water and diethylbenzene.
Formula Weight 134.22
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 1,3-Diethylbenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'