missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethylbarbituric acid, 99% (dry wt.), water <6%, Thermo Scientific Chemicals

Catalog Number p-7052400
29.60 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
769-42-6
C6H8N2O3
156.141
MFCD00006675
VVSASNKOFCZVES-UHFFFAOYSA-N
1,3-dimethylbarbituric acid, n,n'-dimethylbarbituric acid, 2,4,6 1h,3h,5h-pyrimidinetrione, 1,3-dimethyl, 1,3-dimethylbarbituricacid, barbituric acid, 1,3-dimethyl, 1,3-dimethylpyrimidine-2,4,6 1h,3h,5h-trione, 1,3-dimethyl barbituric acid, 1,3-dimethyl-2,4,6 1h,3h,5h-pyrimidinetrione, 1,3-dimethyl-2,4,6 1h,3h,5h pyrimidinetrione, 1,3-dimethyl-1,3,5-trihydropyrimidine-2,4,6-trione
69860
1,3-dimethyl-1,3-diazinane-2,4,6-trione
CN1C(=O)CC(=O)N(C1=O)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
769-42-6
C6H8N2O3
156.141
MFCD00006675
VVSASNKOFCZVES-UHFFFAOYSA-N
1,3-dimethylbarbituric acid, n,n'-dimethylbarbituric acid, 2,4,6 1h,3h,5h-pyrimidinetrione, 1,3-dimethyl, 1,3-dimethylbarbituricacid, barbituric acid, 1,3-dimethyl, 1,3-dimethylpyrimidine-2,4,6 1h,3h,5h-trione, 1,3-dimethyl barbituric acid, 1,3-dimethyl-2,4,6 1h,3h,5h-pyrimidinetrione, 1,3-dimethyl-2,4,6 1h,3h,5h pyrimidinetrione, 1,3-dimethyl-1,3,5-trihydropyrimidine-2,4,6-trione
69860
1,3-dimethyl-1,3-diazinane-2,4,6-trione
CN1C(=O)CC(=O)N(C1=O)C

1,3-Dimethylbarbituric acid is used as a catalyst in the Knoevenagel condensation of a series of aromatic aldehydes. It is also used in the synthesis of 5-aryl-6-(alkyl- or aryl-amino)-1,3-dimethylfuro [2,3-d]pyrimidine derivatives and enantioselective synthesis of isochromene pyrimidinedione derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 121°C to 123°C
Beilstein 139810
Formula Weight 156.14
Percent Purity 99%
Concentration or Composition (by Analyte or Components) water <6%
Quantity 50 g
Physical Form Crystalline Powder
Assay (dry weight)
Chemical Name or Material 1,3-Dimethylbarbituric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'