Learn More
1-(3-Fluorophenyl)pyrrole, 98%, Thermo Scientific™
Product Code. 15486157
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 15486157
Brand: Thermo Scientific Alfa Aesar H33348.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 53871-27-5
Molecular Formula C10H8FN
Molecular Weight (g/mol) 161.18
MDL Number MFCD02665249
InChI Key MMILNBLSOHVLCJ-UHFFFAOYSA-N
Synonym 1-3-fluorophenyl pyrrole, 1-3-fluorophenyl-1h-pyrrole
PubChem CID 4763720
IUPAC Name 1-(3-fluorophenyl)pyrrole
SMILES FC1=CC=CC(=C1)N1C=CC=C1
Specifications
Boiling Point 54°C (0.15 mmHg)
Refractive Index 1.578
Quantity 5 g
Formula Weight 161.18
Percent Purity 98%
Chemical Name or Material 1-(3-Fluorophenyl)pyrrole