missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(1-Piperazinyl)phenyl]ethanone, TRC

Product Code. 30414726
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30414726 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30414726

Brand: Toronto Research Chemicals TRCP4812401G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[4-(1-Piperazinyl)phenyl]ethanone
CAS 51639-48-6
Molecular Formula C12H16N2O
Quantity 1 g
Synonym 4'-Piperazinoacetophenone
InChI Formula InChI=1S/C12H16N2O/c1-10(15)11-2-4-12(5-3-11)14-8-6-13-7-9-14/h2-5,13H,6-9H2,1H3
SMILES CC(=O)c1ccc(cc1)N2CCNCC2
IUPAC Name 1-(4-piperazin-1-ylphenyl)ethanone
Molecular Weight (g/mol) 204.27
Formula Weight 204.1263
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.