missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 1.5 mL Polypropylene Microcentrifuge Tubes

Product Code. 11759502
Click to view available options
Color:
Blue
Green
Yellow
Closure Type:
Cap
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 11759502

Brand: Greiner Bio-One 616275

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

For single use only. Greiner Bio-One Cell 1.5mL Polypropylene Microcentrifuge Tubes are free of heavy metal.

  • Material: Polypropylene
  • For single use only
  • Available in different colors
  • Free of detectable DNase/RNase, human DNA and pyrogens
  • Suitable for Eppendorf centrifuges and systems
TRUSTED_SUSTAINABILITY

Specifications

Color Green
Material Polypropylene
Volume (Metric) 1.5mL
Sterility Non-sterile
Graduated Yes
Closure Type Cap
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.