missing translation for 'onlineSavingsMsg'
Learn More

1,7-Octadiene, 98.5%

Product Code. p-3758581 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 10366770

Brand: Thermo Scientific Acros 129351000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3710-30-3 | C8H14 | 110.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3710-30-3
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00008668
InChI Key XWJBRBSPAODJER-UHFFFAOYSA-N
Synonym 1,7-octadiene, .alpha.,.omega.-octadiene, unii-n4h29t34j2, octa-1,7-dien, acmc-209iqt, 4-01-00-01038 beilstein handbook reference, wln: 1u6u1
PubChem CID 19460
IUPAC Name octa-1,7-diene
SMILES C=CCCCCC=C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -70.0°C
Color Colorless to Yellow
Density 0.7400g/mL
Boiling Point 114.0°C to 121.0°C
Flash Point 9°C
Infrared Spectrum Authentic
Assay Percent Range 98.50%
Packaging Glass bottle
Linear Formula H2C=CH(CH2)4CH=CH2
Refractive Index 1.4210 to 1.4230
Quantity 100 mL
UN Number 2309
Specific Gravity 0.74
Solubility Information Solubility in water: 0.08g/L (20°C). Other solubilities: miscible with methanol, cyclohexane, benzene,, isopropanol and petroleumether
Viscosity 0.5 mPa.s (20°C)
Formula Weight 110.2
Percent Purity 98.5%
Physical Form Liquid
Chemical Name or Material 1, 7-Octadiene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.