missing translation for 'onlineSavingsMsg'
Learn More

1,8-Bis(dimethylamino)naphthalene, 99%, Thermo Scientific Chemicals

Catalog Number p-3523265
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
20734-58-1
C14H18N2
214.31
MFCD00003920
GJFNRSDCSTVPCJ-UHFFFAOYSA-N
1,8-bis dimethylamino naphthalene, n1,n1,n8,n8-tetramethylnaphthalene-1,8-diamine, proton sponge, 1,8-bis dimethylamino naphtalene, n,n,n',n'-tetramethyl-1,8-naphthalenediamine, proton-sponge, 1,8-naphthalenediamine, n,n,n',n'-tetramethyl, unii-6s79d2p9h8, 1,8-bis-dimethylamino naphthalene, n,n,n',n'-tetramethylnaphthalene-1,8-diamine
88675
1-N,1-N,8-N,8-N-tetramethylnaphthalene-1,8-diamine
CN(C)C1=CC=CC2=C1C(=CC=C2)N(C)C
This item is not returnable. View return policy
20734-58-1
C14H18N2
214.31
MFCD00003920
GJFNRSDCSTVPCJ-UHFFFAOYSA-N
1,8-bis dimethylamino naphthalene, n1,n1,n8,n8-tetramethylnaphthalene-1,8-diamine, proton sponge, 1,8-bis dimethylamino naphtalene, n,n,n',n'-tetramethyl-1,8-naphthalenediamine, proton-sponge, 1,8-naphthalenediamine, n,n,n',n'-tetramethyl, unii-6s79d2p9h8, 1,8-bis-dimethylamino naphthalene, n,n,n',n'-tetramethylnaphthalene-1,8-diamine
88675
1-N,1-N,8-N,8-N-tetramethylnaphthalene-1,8-diamine
CN(C)C1=CC=CC2=C1C(=CC=C2)N(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 46°C to 50°C
Color Brown-Pink
Boiling Point 144°C to 145°C (5.0 hPa)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Fieser 03,22; 04,35; 06,50; 13,293
Merck Index 15, 3432
Solubility Information Solubility in water: insoluble
Formula Weight 214.31
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1, 8-Bis(dimethylamino)naphthalene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,8-Bis(dimethylamino)naphtalene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'