missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylpiperazine, 98%

Product Code. 10761234 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10761234 100 g Glass bottle 100g
10205921 5 g Glass bottle 5g
10113211 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10761234 Supplier Thermo Scientific Acros Supplier No. 346831000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 13889-98-0 | C6H12N2O | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13889-98-0
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.17
MDL Number MFCD00058676
InChI Key PKDPUENCROCRCH-UHFFFAOYSA-N
Synonym 1-acetylpiperazine, n-acetylpiperazine, 1-piperazin-1-yl ethanone, 1-piperazin-1-yl ethan-1-one, n-acetyl piperazine, acetylpiperazine, 1-acetyl piperazine, piperazine, 1-acetyl, 1-piperazin-1-yl-ethanone, 1-piperazin-1-yl ethanon
PubChem CID 83795
IUPAC Name 1-piperazin-1-ylethanone
SMILES CC(=O)N1CCNCC1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 31°C to 34°C
Color Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Beilstein 23,IV,201
Formula Weight 128.17
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 1-Acetylpiperazine
Show More Show Less
compliance-icons
Product Identifier
  • 1-Acetylpiperazine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.