Learn More
1-Adamantanemethanol, 98%
Description
1-Adamantanemethanol was used in the synthesis of axially disubstituted silicon(IV) phthalocyanines. It is used as a reagent in the synthesis of adamantane and noradamantane based histone deacetylase (HDAC) inhibitors for the treatment of cancer. It is also used as a reagent in the synthesis of (1-adamantyl)methyl glycidyl ether, a versatile building block for living polymerization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Adamantanemethanol was used in the synthesis of axially disubstituted silicon(IV) phthalocyanines. It is used as a reagent in the synthesis of adamantane and noradamantane based histone deacetylase (HDAC) inhibitors for the treatment of cancer. It is also used as a reagent in the synthesis of (1-adamantyl)methyl glycidyl ether, a versatile building block for living polymerization.
Solubility
Insoluble in water. Solubility in methanol (almost transparency).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 115°C to 118°C |
| Quantity | 5 g |
| Beilstein | 1853339 |
| Solubility Information | Insoluble in water. Solubility in methanol (almost transparency). |
| Formula Weight | 166.26 |
| Percent Purity | 98% |
| Chemical Name or Material | 1-Adamantanemethanol |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.