missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanemethanol, 99%, Thermo Scientific Chemicals

Catalog Number p-50427
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
770-71-8
C11H18O
166.26
MFCD00074751
MDVGOOIANLZFCP-UHFFFAOYSA-N
1-adamantanemethanol, adamantan-1-ylmethanol, 1-adamantane methanol, 1-hydroxymethyl adamantane, adamantane-1-methanol, adamantan-1-yl-methanol, 1-hydroxymethyladamantane, tricyclo 3.3.1.1'3,7 dec-1-ylmethanol, adamantan-1-yl methanol, tricyclo 3.3.1.13,7 decanemethanol
64556
1-adamantylmethanol
C1C2CC3CC1CC(C2)(C3)CO
This item is not returnable. View return policy
770-71-8
C11H18O
166.26
MFCD00074751
MDVGOOIANLZFCP-UHFFFAOYSA-N
1-adamantanemethanol, adamantan-1-ylmethanol, 1-adamantane methanol, 1-hydroxymethyl adamantane, adamantane-1-methanol, adamantan-1-yl-methanol, 1-hydroxymethyladamantane, tricyclo 3.3.1.1'3,7 dec-1-ylmethanol, adamantan-1-yl methanol, tricyclo 3.3.1.13,7 decanemethanol
64556
1-adamantylmethanol
C1C2CC3CC1CC(C2)(C3)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 115.0°C to 118.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 2.5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in hexane and ethyl alcohol
Formula Weight 166.26
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1-Adamantanemethanol, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'