missing translation for 'onlineSavingsMsg'
Learn More

1-Allylpyrazole, 97%, ACROS Organics™

Brand:  Acros Organics 456520050

Additional Details : CAS : 35852-74-5 Weight : 0.00500kg


Quantity 5g

 View more versions of this product

Product Code. 15412172

  • 347.00€ / 5g


Estimated Shipment
27-04-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

35852-74-5
108.14
1-allyl-1h-pyrazole, 1-prop-2-en-1-yl-1h-pyrazole, n-allylpyrazole, pyrazole1allyl
1-prop-2-enylpyrazole
C6H8N2
BPPPRRLOPGKVMQ-UHFFFAOYSA-N
3489293
C=CCN1C=CC=N1

Specifications

1-Allylpyrazole
157.0°C to 158.0°C
35852-74-5
C6H8N2
BPPPRRLOPGKVMQ-UHFFFAOYSA-N
1-prop-2-enylpyrazole
3489293
97%
Authentic
5g
96% min. (GC)
1-allyl-1h-pyrazole, 1-prop-2-en-1-yl-1h-pyrazole, n-allylpyrazole, pyrazole1allyl
C=CCN1C=CC=N1
108.14
108.14
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1-Allylpyrazole

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents