missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-methylpiperazine hydrochloride, 98+%, Thermo Scientific™

Catalog Number p-7030537
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1217863-48-3
C12H19ClN2
226.748
MFCD09842143
OTGMFHMWEOEDNN-UHFFFAOYSA-N
1-benzyl-3-methylpiperazine hydrochloride
53442931
1-benzyl-3-methylpiperazine;hydrochloride
CC1CN(CCN1)CC2=CC=CC=C2.Cl
This item is not returnable. View return policy
1217863-48-3
C12H19ClN2
226.748
MFCD09842143
OTGMFHMWEOEDNN-UHFFFAOYSA-N
1-benzyl-3-methylpiperazine hydrochloride
53442931
1-benzyl-3-methylpiperazine;hydrochloride
CC1CN(CCN1)CC2=CC=CC=C2.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 226.75
Percent Purity ≥98%
Chemical Name or Material 1-Benzyl-3-methylpiperazine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'