missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-N-phenylpiperidin-4-amine, Mikromol™

Product Code. 17921106
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17921106 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17921106 Supplier Mikromol Supplier No. MM0528.150025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-Benzyl-N-phenylpiperidin-4-amine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Phenyl-1-(phenylmethyl)-4-piperidinamine
CAS 1155-56-2
Analyte or Component Names N-Phenyl-1-(phenylmethyl)-4-piperidinamine
Shipping Condition Room Temperature
Molecular Formula C18 H22 N2
Certifications/Compliance ISO 9001
Synonym 4-Piperidinamine, N-phenyl-1-(phenylmethyl)-, 4-anilino-1-benzyl- (7CI,8CI), N-Phenyl-1-(phenylmethyl)-4-piperidinamine, 1-Benzyl-4-(phenylamino)piperidine, 1-Benzyl-4-anilinopiperidine, 4-Anilino-1-benzylpiperidine, NSC 76613, 4-Anilino-1-benzylpiperidine, Fentanyl Related Compound D, Fentanyl Citrate Impurity G, Fentanyl Impurity J, Fentanyl Impurity G
InChI Formula InChI=1S/C18H22N2/c1-3-7-16(8-4-1)15-20-13-11-18(12-14-20)19-17-9-5-2-6-10-17/h1-10,18-19H,11-15H2
SMILES C(N1CCC(CC1)Nc2ccccc2)c3ccccc3
IUPAC Name 1-benzyl-N-phenylpiperidin-4-amine
Molecular Weight (g/mol) 266.38
Quantity 25 mg
Formula Weight 266.1783 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.