missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-3-(aminomethyl)azetidine, 95%, ACROS Organics™

Brand:  Acros Organics 451190010

Additional Details : CAS : 325775-44-8 Weight : 0.01000kg


Quantity 1g

 View more versions of this product

Product Code. 15279626

  • 274.40€ / 1g


Estimated Shipment
31-07-2020
Add to basket


Specifications

Chemical Identifiers

325775-44-8
186.25
1-boc-3-aminomethyl azetidine, tert-butyl 3-aminomethyl azetidine-1-carboxylate, 1-boc-3-aminomethylazetidine, 3-aminomethyl-1-n-boc-azetidine, 1-n-boc-3-aminomethylazetidine, 3-aminomethyl-azetidine-1-carboxylic acid tert-butyl ester, 1-boc-3-aminomethyl-azetidine, 3-aminomethyl azetidine, n1-boc protected, tert-butyl 3-aminomethyl azetidinecarboxylate, 1-azetidinecarboxylic acid, 3-aminomethyl-, 1,1-dimethylethyl ester
tert-butyl 3-(aminomethyl)azetidine-1-carboxylate
C9H18N2O2
XSJPKMUFBHSIRA-UHFFFAOYSA-N
10679125
CC(C)(C)OC(=O)N1CC(C1)CN

Specifications

1-BOC-3-(aminomethyl)azetidine
Glass Bottle
325775-44-8
C9H18N2O2
XSJPKMUFBHSIRA-UHFFFAOYSA-N
tert-butyl 3-(aminomethyl)azetidine-1-carboxylate
10679125
95%
>110°C
1g
94% min. (GC)
1-boc-3-aminomethyl azetidine, tert-butyl 3-aminomethyl azetidine-1-carboxylate, 1-boc-3-aminomethylazetidine, 3-aminomethyl-1-n-boc-azetidine, 1-n-boc-3-aminomethylazetidine, 3-aminomethyl-azetidine-1-carboxylic acid tert-butyl ester, 1-boc-3-aminomethyl-azetidine, 3-aminomethyl azetidine, n1-boc protected, tert-butyl 3-aminomethyl azetidinecarboxylate, 1-azetidinecarboxylic acid, 3-aminomethyl-, 1,1-dimethylethyl ester
CC(C)(C)OC(=O)N1CC(C1)CN
186.25
186.25
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1-Boc-3-(aminomethyl)azetidine, 95%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Documents