missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Boc-4-(2-chloro-6-nitrophenyl)piperazine, 97%

Brand:  Alfa Aesar™ H63127.03

Additional Details : CAS : 400803-05-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409648

  • 82.68€ / 1g


Estimated Shipment
14-05-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

400803-05-6
tert-Butyl 4-(2-chloro-6-nitrophenyl)-piperazine-1-carboxylate; 4-(2-Chloro-6-nitrophenyl)-piperazine-1-carboxylic acid tert-butyl ester

Specifications

1-Boc-4-(2-chloro-6-nitrophenyl)piperazine
400803-05-6
tert-Butyl 4-(2-chloro-6-nitrophenyl)-piperazine-1-carboxylate; 4-(2-Chloro-6-nitrophenyl)-piperazine-1-carboxylic acid tert-butyl ester
Solid
1g
14092196
341.79
97%
Videos
SDS Documents