Learn More
Alfa Aesar™ 1-Boc-4-(2-Fluoro-6-nitrophenyl)piperazine, 97%
Brand: Alfa Aesar™ H63668.03
Additional Details : CAS : 1233958-36-5 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
1233958-36-5 | |
325.34 | |
GWHUVEIGDXIWOY-UHFFFAOYSA-N | |
53255503 | |
CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=CC=C2F)[N+](=O)[O-] |
C15H20FN3O4 | |
MFCD14155993 | |
tert-butyl 4-2-fluoro-6-nitrophenyl piperazine-1-carboxylate, 1-boc-4-2-fluoro-6-nitrophenyl piperazine | |
tert-butyl 4-(2-fluoro-6-nitrophenyl)piperazine-1-carboxylate |
Specifications
1-Boc-4-(2-Fluoro-6-nitrophenyl)piperazine | |
1233958-36-5 | |
MFCD14155993 | |
tert-butyl 4-2-fluoro-6-nitrophenyl piperazine-1-carboxylate, 1-boc-4-2-fluoro-6-nitrophenyl piperazine | |
CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=CC=C2F)[N+](=O)[O-] | |
325.34 | |
325.34 | |
97% |
1g | |
C15H20FN3O4 | |
19933459 | |
GWHUVEIGDXIWOY-UHFFFAOYSA-N | |
tert-butyl 4-(2-fluoro-6-nitrophenyl)piperazine-1-carboxylate | |
53255503 | |
Solid |