missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-(4-nitrophenyl)piperazine, 97%, Thermo Scientific™

Catalog Number p-7055116
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
182618-86-6
C15H21N3O4
307.35
MFCD02187326
MGYCIJUTYLUYJM-UHFFFAOYSA-N
1-boc-4-4-nitrophenyl piperazine, tert-butyl 4-4-nitrophenyl piperazine-1-carboxylate, 4-4-nitrophenyl piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 4-4-nitrophenyl-, 1,1-dimethylethyl ester, tert-butyl 4-4-nitrophenyl piperazinecarboxylate, acmc-1c6wn, 1-n-boc-4-4-nitrophenyl piperazine, 1-4-boc-piperazin-1-yl-4-nitrobenzene, 1-t-butoxycarbonyl4-4-nitrophenyl piperazine, 1-t-butoxycarbonyl-4-4-nitrophenyl piperazine
3380696
tert-butyl 4-(4-nitrophenyl)piperazine-1-carboxylate
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]
This item is not returnable. View return policy
182618-86-6
C15H21N3O4
307.35
MFCD02187326
MGYCIJUTYLUYJM-UHFFFAOYSA-N
1-boc-4-4-nitrophenyl piperazine, tert-butyl 4-4-nitrophenyl piperazine-1-carboxylate, 4-4-nitrophenyl piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 4-4-nitrophenyl-, 1,1-dimethylethyl ester, tert-butyl 4-4-nitrophenyl piperazinecarboxylate, acmc-1c6wn, 1-n-boc-4-4-nitrophenyl piperazine, 1-4-boc-piperazin-1-yl-4-nitrobenzene, 1-t-butoxycarbonyl4-4-nitrophenyl piperazine, 1-t-butoxycarbonyl-4-4-nitrophenyl piperazine
3380696
tert-butyl 4-(4-nitrophenyl)piperazine-1-carboxylate
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 307.34
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Boc-4-(4-nitrophenyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'