missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Boc-4,5-dicyano-2-(2-pyridyl)imidazole, 97%

Brand:  Alfa Aesar™ H54235.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15410578

  • 1045.00€ / 5g


Estimated Shipment
12-05-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

1347815-01-3
295.302
OOHIBQVWUXPAKV-UHFFFAOYSA-N
73995706
CC(C)(C)OC(=O)N1C(=C(N=C1C2=CC=CC=N2)C#N)C#N
C15H13N5O2
MFCD20265212
1-boc-4,5-dicyano-2-2-pyridyl imidazole, tert-butyl 4,5-dicyano-2-pyridin-2-yl imidazole-1-carboxylate
tert-butyl 4,5-dicyano-2-pyridin-2-ylimidazole-1-carboxylate

Specifications

1-Boc-4,5-dicyano-2-(2-pyridyl)imidazole
1347815-01-3
MFCD20265212
OOHIBQVWUXPAKV-UHFFFAOYSA-N
tert-butyl 4,5-dicyano-2-pyridin-2-ylimidazole-1-carboxylate
73995706
97%
5g
C15H13N5O2
1-boc-4,5-dicyano-2-2-pyridyl imidazole, tert-butyl 4,5-dicyano-2-pyridin-2-yl imidazole-1-carboxylate
CC(C)(C)OC(=O)N1C(=C(N=C1C2=CC=CC=N2)C#N)C#N
295.302
295.3
Videos
SDS Documents