missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-iodobenzene, 98+%, stab. with copper, Thermo Scientific Chemicals

Catalog Number p-7023455
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
583-55-1
C6H4BrI
282.906
MFCD00001030
OIRHKGBNGGSCGS-UHFFFAOYSA-N
2-bromoiodobenzene, o-bromoiodobenzene, 2-bromo-iodobenzene, benzene, 1-bromo-2-iodo, bromoiodobenzene, 1-bromo-2-iodo-benzene, attercop-chm at127403, chemwish ic03500, iodobromobenzene, 2-iodobromobenzene
11415
1-bromo-2-iodobenzene
C1=CC=C(C(=C1)Br)I
This item is not returnable. View return policy
583-55-1
C6H4BrI
282.906
MFCD00001030
OIRHKGBNGGSCGS-UHFFFAOYSA-N
2-bromoiodobenzene, o-bromoiodobenzene, 2-bromo-iodobenzene, benzene, 1-bromo-2-iodo, bromoiodobenzene, 1-bromo-2-iodo-benzene, attercop-chm at127403, chemwish ic03500, iodobromobenzene, 2-iodobromobenzene
11415
1-bromo-2-iodobenzene
C1=CC=C(C(=C1)Br)I

2-Bromoiodobenzene is used in the synthesis of diarylamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoiodobenzene is used in the synthesis of diarylamines.

Solubility
Not miscible or difficult to mix in water.

Notes
Light Sensitive. Protect from light. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 9°C to 10°C
Density 2.256
Boiling Point 257°C
Flash Point >110°C (230°F)
Refractive Index 1.6645
Quantity 5 g
Beilstein 1855309
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 282.91
Percent Purity ≥98%
Chemical Name or Material 1-Bromo-2-iodobenzene, stabilized with copper
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'