missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-fluoro-2-(2,2,2-trifluoroethyl)benzene, 95%, Thermo Scientific™

Catalog Number 15408455
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1186194-80-8
C8H5BrF4
257.026
MFCD11226691
VDKXBQCWYHXJNB-UHFFFAOYSA-N
1-bromo-4-fluoro-2-2,2,2-trifluoroethyl benzene
45790615
1-bromo-4-fluoro-2-(2,2,2-trifluoroethyl)benzene
C1=CC(=C(C=C1F)CC(F)(F)F)Br
This item is not returnable. View return policy
1186194-80-8
C8H5BrF4
257.026
MFCD11226691
VDKXBQCWYHXJNB-UHFFFAOYSA-N
1-bromo-4-fluoro-2-2,2,2-trifluoroethyl benzene
45790615
1-bromo-4-fluoro-2-(2,2,2-trifluoroethyl)benzene
C1=CC(=C(C=C1F)CC(F)(F)F)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 76°C (15 mmHg)
Quantity 1 g
Formula Weight 257.02
Percent Purity 95%
Chemical Name or Material 1-Bromo-4-fluoro-2-(2,2,2-trifluoroethyl)benzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'