missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butanol-d, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 1-Butanol-d |
| CAS | 4712-38-3 |
| Molecular Formula | C4 D H9 O |
| Quantity | 250 mg |
| Synonym | 1-Butanol-d (9CI), Butyl alcohol-d (6CI, 7CI, 8CI), 1-Deuteriooxybutane, Butanol-O-d, n-Butanol-O-d, 1-Butanol-O-D, n-Butyl alcohol-OD |
| InChI Formula | InChI=1S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3/i5D |
| SMILES | [2H]OCCCC |
| IUPAC Name | 1-deuteriooxybutane |
| Molecular Weight (g/mol) | 75.128 |
| Formula Weight | 75.079 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction