missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butene-3,4-diol, TRC
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
Specifications
Specifications
| Chemical Name or Material | 1-Butene-3,4-diol |
| CAS | 497-06-3 |
| Molecular Formula | C4H8O2 |
| Quantity | 100 mg |
| Synonym | 3,4-Dihydroxy-1-butene, Erythrol |
| InChI Formula | InChI=1S/C4H8O2/c1-2-4(6)3-5/h2,4-6H,1,3H2 |
| SMILES | OCC(O)C=C |
| IUPAC Name | but-3-ene-1,2-diol |
| Molecular Weight (g/mol) | 88.11 |
| Formula Weight | 88.0524 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction