missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-3-[1-(4-chlorophenyl)ethyl]benzene, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
1-Chloro-3-[1-(4-chlorophenyl)ethyl]benzene
Specifications
Specifications
| Chemical Name or Material | 1-Chloro-3-[1-(4-chlorophenyl)ethyl]benzene |
| CAS | 77008-64-1 |
| Analyte or Component Names | 1-Chloro-3-[1-(4-chlorophenyl)ethyl]benzene |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C14 H12 Cl2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-Chloro-3-[1-(4-chlorophenyl)ethyl]benzene |
| InChI Formula | InChI=1S/C14H12Cl2/c1-10(11-5-7-13(15)8-6-11)12-3-2-4-14(16)9-12/h2-10H,1H3 |
| SMILES | CC(c1ccc(Cl)cc1)c2cccc(Cl)c2 |
| IUPAC Name | 1-chloro-3-[1-(4-chlorophenyl)ethyl]benzene |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction