missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-6-hydroxyhexane, 95%, Thermo Scientific Chemicals

Catalog Number p-3735985
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
2009-83-8
C6H13ClO
136.62
MFCD00002984
JNTPTNNCGDAGEJ-UHFFFAOYSA-N
6-chloro-1-hexanol, 6-chlorohexanol, 1-chloro-6-hydroxyhexane, 1-hexanol, 6-chloro, hexamethylene chlorohydrin, 1-chloro-6-hexanol, omega-chlorohexanol, unii-t2hit6n5uf, 6-chloro-hexanol, t2hit6n5uf
74828
6-chlorohexan-1-ol
C(CCCCl)CCO
This item is not returnable. View return policy
2009-83-8
C6H13ClO
136.62
MFCD00002984
JNTPTNNCGDAGEJ-UHFFFAOYSA-N
6-chloro-1-hexanol, 6-chlorohexanol, 1-chloro-6-hydroxyhexane, 1-hexanol, 6-chloro, hexamethylene chlorohydrin, 1-chloro-6-hexanol, omega-chlorohexanol, unii-t2hit6n5uf, 6-chloro-hexanol, t2hit6n5uf
74828
6-chlorohexan-1-ol
C(CCCCl)CCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0300g/mL
Boiling Point 108.0°C to 112.0°C (14.0 mmHg)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula Cl(CH2)6OH
Refractive Index 1.4547 to 1.4567
Quantity 25 g
Specific Gravity 1.03
Formula Weight 136.62
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-chloro-6-hydroxyhexane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'