Learn More
1-(Cyclopropylcarbonyl)piperazine, 97%, Thermo Scientific™
Catalog Number p-7077488
Click to view available options
Quantity:
1 g
5 g
59878-57-8
C8H15N2O
155.22
MFCD06369645
KIALFUYSJAAJSU-UHFFFAOYSA-O
1-cyclopropylcarbonyl piperazine, cyclopropyl piperazin-1-yl methanone, 1-cyclopropanecarbonylpiperazine, cyclopropyl-piperazin-1-yl-methanone, 1-cyclopropanecarbonyl piperazine, piperazine, 1-cyclopropylcarbonyl, piperazinocyclopropyl ketone, cyclopropyl piperazinyl ketone, 1-cyclopropylcarbonylpiperazine, 4-cyclopropanecarbonyl-piperazine
2064235
cyclopropyl(piperazin-1-yl)methanone
O=C(C1CC1)N1CC[NH2+]CC1
59878-57-8
C8H15N2O
155.22
MFCD06369645
KIALFUYSJAAJSU-UHFFFAOYSA-O
1-cyclopropylcarbonyl piperazine, cyclopropyl piperazin-1-yl methanone, 1-cyclopropanecarbonylpiperazine, cyclopropyl-piperazin-1-yl-methanone, 1-cyclopropanecarbonyl piperazine, piperazine, 1-cyclopropylcarbonyl, piperazinocyclopropyl ketone, cyclopropyl piperazinyl ketone, 1-cyclopropylcarbonylpiperazine, 4-cyclopropanecarbonyl-piperazine
2064235
cyclopropyl(piperazin-1-yl)methanone
O=C(C1CC1)N1CC[NH2+]CC1
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 154.21
Percent Purity 97%
Chemical Name or Material 1-(Cyclopropylcarbonyl)piperazine
compliance-icons compliance-icons
Product Identifier
  • 1-(Cyclopropylcarbonyl)piperazine, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.