missing translation for 'onlineSavingsMsg'
Learn More

1-Docosanol, TRC

Product Code. 30377056
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Product Code. Quantity
30377056 1 g
30372005 50 g
30384551 10 g
3 options
This item is not returnable. View return policy

Product Code. 30377056

Brand: Toronto Research Chemicals TRCD6788551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 661-19-8
Formula Weight 326.3549
InChI Formula InChI=1S/C22H46O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23/h23H,2-22H2,1H3
IUPAC Name docosan-1-ol
Molecular Formula C22 H46 O
Molecular Weight (g/mol) 326.6
SMILES CCCCCCCCCCCCCCCCCCCCCCO
Synonym 1-Docosanol, 1-Docosonol, Abreva, Behenic alcohol, Behenyl 80 Alcohol, Behenyl alc. 80, Behenyl alcohol, Conol 2265, Conol 2280, Docosanol, Docosyl alcohol, Hainol 22S, IK 2, IK 2 (alcohol), Kalcohl 220, Kalcohl 22080, Kalcol 22080, Lanette 22, NAA 422, NSC 8407, Nacol 22-97, Nacol 22-98, Nacol C 22, Stenol 1822, Stenol 1822A, Tadenan, Toho BH 65, n-Docosanol
Chemical Name or Material n-Docosanol

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.