missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecyne, 98%, Thermo Scientific Chemicals

Catalog Number p-3757788
34.66 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
765-03-7
C12H22
166.31
MFCD00008960
ZVDBUOGYYYNMQI-UHFFFAOYSA-N
1-dodecyne, dodecyne, dodec-1-in, 1dodecyne, dodec-1-yn, 1-dodecyne 5g
69821
dodec-1-yne
CCCCCCCCCCC#C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
765-03-7
C12H22
166.31
MFCD00008960
ZVDBUOGYYYNMQI-UHFFFAOYSA-N
1-dodecyne, dodecyne, dodec-1-in, 1dodecyne, dodec-1-yn, 1-dodecyne 5g
69821
dodec-1-yne
CCCCCCCCCCC#C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -19°C
Color Colorless to Yellow
Density 0.7700g/mL
Boiling Point 215°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)9C∽CH
Refractive Index 1.4330 to 1.435
Quantity 5 mL
Beilstein 01, 261
Specific Gravity 0.77
Formula Weight 166.31
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Dodecyne
Show More Show Less
compliance-icons
Product Identifier
  • 1-Dodecyne
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'