missing translation for 'onlineSavingsMsg'
Learn More

1-Eicosanethiol, TRC

Product Code. 30406991
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
30406991 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 30406991

Brand: Toronto Research Chemicals TRCE975138100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Eicosanethiol
CAS 13373-97-2
Molecular Formula C20H42S
Quantity 100 mg
Synonym n-Eicosylmercaptan
InChI Formula InChI=1S/C20H42S/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21/h21H,2-20H2,1H3
SMILES CCCCCCCCCCCCCCCCCCCCS
IUPAC Name icosane-1-thiol
Molecular Weight (g/mol) 314.61
Formula Weight 314.3007
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.