missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-3-methylimidazolium iodide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
50 g
Description
It is used as OLED materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as OLED materials.
Solubility
Soluble in water, , dichloromethane, ethanol, acetonitrile.
Notes
Moisture Sensitive. Keep away from heat and humidity. Incompatible with strong oxidizing agents.
It is used as OLED materials.
Solubility
Soluble in water, , dichloromethane, ethanol, acetonitrile.
Notes
Moisture Sensitive. Keep away from heat and humidity. Incompatible with strong oxidizing agents.
Specifications
Color | White to Yellow |
Quantity | 5 g |
Beilstein | 5160333 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in water,,dichloromethane,ethanol,acetonitrile. |
Formula Weight | 238.07 |
Percent Purity | 97% |
Physical Form | Powder |
Chemical Name or Material | 1-Ethyl-3-methylimidazolium iodide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'