missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Fluoro-2,4-dimethoxybenzene, 97%

Brand:  Alfa Aesar™ H63528.03

Additional Details : CAS : 17715-70-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15410269

  • 88.20€ / 1g


Estimated Shipment
11-05-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

17715-70-7
156.156
QLJNEPOEZGFNEA-UHFFFAOYSA-N
2758413
COC1=CC(=C(C=C1)F)OC
C8H9FO2
MFCD00042265
2,4-dimethoxy-1-fluorobenzene, 2,4-dimethoxyfluorobenzene, 1,3-dimethoxy-4-fluorobenzene, 4-fluoro-3-methoxyanisole, benzene, 1-fluoro-2,4-dimethoxy, 2,4-dimethoxy-1-fluoro-benzene, acmc-1c4p9, 1-fluoro-2,4-dimethoxy-benzene, qljnepoezgfnea-uhfffaoysa, benzene,1-fluoro-2,4-dimethoxy
1-fluoro-2,4-dimethoxybenzene

Specifications

1-Fluoro-2,4-dimethoxybenzene
105°C (221°F)
1g
C8H9FO2
2,4-dimethoxy-1-fluorobenzene, 2,4-dimethoxyfluorobenzene, 1,3-dimethoxy-4-fluorobenzene, 4-fluoro-3-methoxyanisole, benzene, 1-fluoro-2,4-dimethoxy, 2,4-dimethoxy-1-fluoro-benzene, acmc-1c4p9, 1-fluoro-2,4-dimethoxy-benzene, qljnepoezgfnea-uhfffaoysa, benzene,1-fluoro-2,4-dimethoxy
COC1=CC(=C(C=C1)F)OC
156.156
156.15
97%
1.188 g/mL
Yellow
17715-70-7
MFCD00042265
QLJNEPOEZGFNEA-UHFFFAOYSA-N
1-fluoro-2,4-dimethoxybenzene
2758413
Liquid
Videos
SDS Documents