missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hexanesulfonic Acid Sodium Salt, HPLC Grade, J.T. Baker™
256.50 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
25 g
Packaging:
Wide Mouth Amber Glass Bottle
Description
- For Ion-Pair Chromatography of Basic Compounds
Specifications
Packaging | Wide Mouth Amber Glass Bottle |
Quantity | 25 g |
Specific Gravity | 1L = 0.00kg |
IUPAC Name | sodium;hexane-1-sulfonate |
Formula Weight | 188.22 |
Grade | HPLC |
Chemical Name or Material | 1-Hexanesulfonic Acid Sodium Salt |
Safety and Handling
Product Identifier
- 1-HEXANESULFONIC ACID SODIUM SALT
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : SODIUM 1-HEXANESULFONATE
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'