missing translation for 'onlineSavingsMsg'
Learn More

1-Hexylmagnesium chloride, 1M in MeTHF, Thermo Scientific™

Product Code. 15418057
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 15418057

Brand: Thermo Scientific Alfa Aesar H51167.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 44767-62-6
Molecular Formula C6H13ClMg
Molecular Weight (g/mol) 144.93
MDL Number MFCD06798087
InChI Key ODOOOEXDBNGOOF-UHFFFAOYSA-M
Synonym hexylmagnesium chloride, 1-hexylmagnesium chloride, hexylmagnesium chloride solution, hexyl magnesium chloride, n-hexylmagnesium chloride, chloro hexyl magnesium, odoooexdbngoof-uhfffaoysa-m, 1-hexylmagnesium chloride, 1m in methf, hexylmagnesium chloride2.0m solution, hexylmagnesium chloride solution, 2.0 m in thf
PubChem CID 10877247
SMILES CCCCCC[Mg]Cl

Specifications

Linear Formula CH3(CH2)4CH2MgCl
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
IUPAC Name chloro(hexyl)magnesium
Formula Weight 144.93
Concentration or Composition (by Analyte or Components) 1M in MeTHF
Chemical Name or Material 1-Hexylmagnesium chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.