missing translation for 'onlineSavingsMsg'
Learn More

1-Iodopropane, 99%, stabilized, Thermo Scientific Chemicals

Catalog Number p-149126
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
107-08-4
C3H7I
169.99
MFCD00001095
FMKOJHQHASLBPH-UHFFFAOYSA-N
propyl iodide, n-propyl iodide, propane, 1-iodo, propane, iodo, 1-iodo-propane, 1-jodpropan, iodopropane, unii-nd3629ke2k, propyliodide, iodopropanes
33643
1-iodopropane
CC(C)I
This item is not returnable. View return policy
107-08-4
C3H7I
169.99
MFCD00001095
FMKOJHQHASLBPH-UHFFFAOYSA-N
propyl iodide, n-propyl iodide, propane, 1-iodo, propane, iodo, 1-iodo-propane, 1-jodpropan, iodopropane, unii-nd3629ke2k, propyliodide, iodopropanes
33643
1-iodopropane
CC(C)I

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -101.0°C
Color Colorless to Yellow
Density 1.7400g/mL
Boiling Point 101.0°C to 102.0°C
Flash Point 44°C
Infrared Spectrum Authentic
Assay Percent Range 1.5% max. (Sum of all by-products) (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2I
Refractive Index 1.5030 to 1.5050
Quantity 50 mL
Beilstein 01, 113
Merck Index 15, 7974
Specific Gravity 1.74
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol and ether
Stabilizer copper
Formula Weight 169.99
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Iodopropane, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Iodopropane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'