missing translation for 'onlineSavingsMsg'
Learn More

1-Isopropenyl-2-benzimidazolidinone, 98+%, Thermo Scientific Chemicals

Catalog Number p-7047111
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
52099-72-6
C10H10N2O
174.203
MFCD00218253
XFASJWLBXHWUMW-UHFFFAOYSA-N
1-prop-1-en-2-yl-1h-benzo d imidazol-2 3h-one, 1-isopropenyl-2-benzimidazolidinone, 1,3-dihydro-1-1-methylethenyl-2h-benzimidazole-2-one, 1-isopropenylbenzimidazolone, 1,3-dihydro-1-1-methylvinyl-2h-benzimidazol-2-one, 1-prop-1-en-2-yl-2,3-dihydro-1h-1,3-benzodiazol-2-one, 1-isopropenyl-1,3-dihydro-benzoimidazol-2-one, 1-isopropenyl-2-benzimidazolinone, 1-isopropenyl-1,3-dihydro-2h-1,3-benzimidazol-2-one, 1-isopropenyl-2,3-dihydro-1h-benzo d imidazol-2-one
100278
3-prop-1-en-2-yl-1H-benzimidazol-2-one
CC(=C)N1C2=CC=CC=C2NC1=O
This item is not returnable. View return policy
52099-72-6
C10H10N2O
174.203
MFCD00218253
XFASJWLBXHWUMW-UHFFFAOYSA-N
1-prop-1-en-2-yl-1h-benzo d imidazol-2 3h-one, 1-isopropenyl-2-benzimidazolidinone, 1,3-dihydro-1-1-methylethenyl-2h-benzimidazole-2-one, 1-isopropenylbenzimidazolone, 1,3-dihydro-1-1-methylvinyl-2h-benzimidazol-2-one, 1-prop-1-en-2-yl-2,3-dihydro-1h-1,3-benzodiazol-2-one, 1-isopropenyl-1,3-dihydro-benzoimidazol-2-one, 1-isopropenyl-2-benzimidazolinone, 1-isopropenyl-1,3-dihydro-2h-1,3-benzimidazol-2-one, 1-isopropenyl-2,3-dihydro-1h-benzo d imidazol-2-one
100278
3-prop-1-en-2-yl-1H-benzimidazol-2-one
CC(=C)N1C2=CC=CC=C2NC1=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 121°C to 123°C
Quantity 1 g
Beilstein 140587
Formula Weight 174.2
Percent Purity ≥98%
Chemical Name or Material 1-Isopropenyl-2-benzimidazolidinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'