missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indazole-4-boronic acid, 95%, Thermo Scientific™

Catalog Number p-7077473
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1001907-60-3
C8H9BN2O2
175.98
MFCD09870045
MDEHELJMJCXYIU-UHFFFAOYSA-N
1-methyl-1h-indazol-4-yl boronic acid, 1-methylindazole-4-boronic acid, 1-methyl-1h-indazole-4-boronic acid, 1-methylindazol-4-boronic acid, 1-methyl-1h-indazol-4-ylboronic acid, 1-methyl-1h-indazol-4-yl-4-boronic acid, acmc-2097nc, 4-borono-1-methyl-1h-indazole, 1-methyl-4-indazolyl boronic acid, 1-methylindazol-4-yl boronic acid
44118212
CN1N=CC2=C(C=CC=C12)B(O)O
This item is not returnable. View return policy
1001907-60-3
C8H9BN2O2
175.98
MFCD09870045
MDEHELJMJCXYIU-UHFFFAOYSA-N
1-methyl-1h-indazol-4-yl boronic acid, 1-methylindazole-4-boronic acid, 1-methyl-1h-indazole-4-boronic acid, 1-methylindazol-4-boronic acid, 1-methyl-1h-indazol-4-ylboronic acid, 1-methyl-1h-indazol-4-yl-4-boronic acid, acmc-2097nc, 4-borono-1-methyl-1h-indazole, 1-methyl-4-indazolyl boronic acid, 1-methylindazol-4-yl boronic acid
44118212
CN1N=CC2=C(C=CC=C12)B(O)O
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 250 mg
IUPAC Name (1-methyl-1H-indazol-4-yl)boronic acid
Formula Weight 175.98
Percent Purity 95%
Chemical Name or Material 1-Methyl-1H-indazole-4-boronic acid
compliance-icons
Product Identifier
  • 1-Methyl-1H-indazole-4-boronic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'