missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone, Biotech Grade, ≥99.7%, Honeywell™ Burdick & Jackson™

Catalog Number 15655680
Click to view available options
Quantity:
1 L
2 L
2.5 L
4 x 2 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
872-50-4
C5H9NO
99.133
MFCD00003193
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
13387
CHEBI:7307
1-methylpyrrolidin-2-one
CN1CCCC1=O
This item is not returnable. View return policy
872-50-4
C5H9NO
99.133
MFCD00003193
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
13387
CHEBI:7307
1-methylpyrrolidin-2-one
CN1CCCC1=O

Biotech. grade,≥99.7%

  • Honeywell Burdick & Jackson™ (B&J) developed the first high-purity solvents more than 50 years ago
  • As new applications and techniques emerge, Burdick & Jackson continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
  • Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
TRUSTED_SUSTAINABILITY
pH 8.5 to 10 (at 20°C)
Melting Point -24°C
Beilstein 106420
Formula Weight 99.13g/mol
Percent Purity ≥99.7%
Density 1.028g/mL at 25°C
Boiling Point 204°C
Quantity 2 L
Explosion Limit 0.095
Flash Point 91°C (195.8°F)
Ignition Point 518°F
Odor Amine like
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 0.99mmHg (40°C), 0.29mmHg (20°C)
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'