missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylpropyl Ester Methanesulfonic Acid, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 1-Methylpropyl Ester Methanesulfonic Acid |
| CAS | 16156-54-0 |
| Molecular Formula | C5H12O3S |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C5H12O3S/c1-4-5(2)8-9(3,6)7/h5H,4H2,1-3H3 |
| SMILES | CCC(C)OS(=O)(=O)C |
| IUPAC Name | butan-2-yl methanesulfonate |
| Molecular Weight (g/mol) | 152.21 |
| Formula Weight | 152.0507 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction