missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TdB Labs AB 500 kDa FITC-TRITC-dextran for Precise Intracellular and Extracellular pH Mapping
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
- Product Overview: FITC-TRITC-dextran 500 kDa is a dual-labeled dextran designed for precise intracellular and extracellular pH mapping. FITC provides pH-sensitive fluorescence, while TRITC offers a pH-stable reference, enabling accurate ratiometric imaging in live-cell and tissue environments.
- Features:
- Dual Fluorescence: FITC (λex 495 nm / λem 519 nm) and TRITC (λex 555 nm / λem 580 nm).
- Ratiometric Imaging: Enables normalized, concentration-independent pH measurement.
- High Molecular Weight: 500 kDa supports restricted diffusion for localized pH measurements.
- Water-Soluble and Biocompatible: Suitable for dynamic and fixed-cell studies.
- Applications:
- Precise pH mapping in live-cell imaging.
- Monitoring pH gradients in tissue engineering and disease models.
- Dynamic and endpoint ratiometric fluorescence assays.
Specifications
Specifications
| Quantity | 1 mg |
| Product Type | Fluorescent Labeling Reagent |
| Molecular Weight (g/mol) | 500 kDa |
| Label Type | Other Labels or Dyes |
| Color | Green |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction